Learn More
4,5-Dichloro-1,3-benzothiazol-2-amine, TRC
Code produit 30377468
Change view
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30377468 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30377468 Fournisseur Toronto Research Chemicals Code fournisseur TRCD434700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 4,5-Dichloro-1,3-benzothiazol-2-amine
CAS 1849-71-4
Formule moléculaire C7H4Cl2N2S
Quantité 100 mg
Formule InChI InChI=1S/C7H4Cl2N2S/c8-3-1-2-4-6(5(3)9)11-7(10)12-4/h1-2H,(H2,10,11)
SMILES Nc1nc2c(Cl)c(Cl)ccc2s1
Poids moléculaire (g/mol) 219.09
Poids de la formule 217.9472