missing translation for 'onlineSavingsMsg'
Learn More

Ester pinacolique d’acide boronique de 4-(N-[2-(Diméthylamino)éthyl]carbamoyl)benzène, 95 %, Thermo Scientific™

Product Code. 15499685
Change view
Click to view available options
Quantité:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
15499685 5 g 5g
15489685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499685 Supplier Thermo Scientific Alfa Aesar Supplier No. H52270.06
Demander un format en vrac ou personnalisé

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ce produit de la marque Thermo Scientific faisait à l’origine partie de la gamme de produits Alfa Aesar. Certains documents et informations sur l’étiquette peuvent faire référence à la marque héritée. Le code d’article / produit Alfa Aesar d’origine ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832114-11-1
Formule moléculaire C17H27BN2O3
Poids moléculaire (g/mol) 318.224
Numéro MDL MFCD05864307
Clé InChI KTDBEJMIGCLCSU-UHFFFAOYSA-N
Synonyme n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-n',n'-dimethylamino ethyl benzamide-4-boronic acid, pinacol ester, n-2-dimethylamino ethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, n-2-dimethylamino ethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 4-2-dimethylamino ethylcarbamoyl phenylboronic acid, pinacol ester, amtb351, n-2-dimethylaminoethyl-4-4,4,5,5-tetramethyl-1,, 4-2-dimethylamino ethyl carbamoyl phenylboronic acid pinacol ester, 4-n-2-dimethylamino ethyl carbamoyl benzeneboronic acid pinacol ester
CID PubChem 2758503
Nom IUPAC N-[2-(diméthylamino)éthyl]-4-(4,4,5,5-tétraméthyl-1,3,2-dioxaborolane-2-yl)benzamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)NCCN(C)C

Specifications

Nom chimique ou matériau 4-(N-[2-(Dimethylamino)ethyl]carbamoyl)benzeneboronic acid pinacol ester
Quantité 5 g
Poids de la formule 318.22
Pourcentage de pureté 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.