Learn More
MedChemExpress 5(6)-ROX
Code produit 16471158
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Conditionnement:
100mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16471158 5 mg -
16474657 50 mg 50mg
16438748 10 mg -
16428886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16471158 Fournisseur MedChemExpress Code fournisseur HYD0043
MedChemExpress 5(6)-ROX is a nucleic acid fluorescent label which can be used as a reference dye for real-time polymerase chain reaction.
Spécification
Contenu et stockage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 50 mg/mL (93.53 mM; Need ultrasonic)
Cible HBV
Type de produit Dye Reagents
Poids moléculaire 534.6 g/mol
CAS 198978-94-8
Synonyme 5(6)-Carboxy-X-rhodamine