Learn More
5-Chloroquinoline-8-sulfonyl Chloride, TRC
Code produit 30364900
Change view
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30364900 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30364900 Fournisseur Toronto Research Chemicals Code fournisseur TRCC381250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 5-Chloroquinoline-8-sulfonyl Chloride, Hydrochloride
CAS 21121-54-0
Formule moléculaire C9 H5 Cl2 N O2 S . Cl H
Quantité 100 mg
Formule InChI InChI=1S/C9H5Cl2NO2S.ClH/c10-7-3-4-8(15(11,13)14)9-6(7)2-1-5-12-9;/h1-5H;1H
SMILES Cl.Clc1ccc(c2ncccc12)S(=O)(=O)Cl
Nom IUPAC 5-chloroquinoline-8-sulfonyl chloride;hydrochloride
Poids moléculaire (g/mol) 298.57
Poids de la formule 296.9185