Learn More
Mecmesin™ Pince pour tissu 5 KN
Code produit 11963918
Encompass
Modifier l'affichage
Click to view available options
Dimensions:
Large
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Dimensions Conditionnement
11963918 Large 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11963918 Fournisseur Mecmesin™ Code fournisseur 432398
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
The 5 kN Wedge Grip is ideal for tensile testing of flat and rigid materials, including plastics, rubber and light metals
  • Spring-loaded lever ensures the serrated jaws are easy to open for fast securing and release of samples. As load is applied, the self-tightening action of the wedge jaws ensures the sample is held rigidly without slippage.
  • Fitted with a 5-16 UNC threaded hole and grub-screw for fixing to a loadcell/gauge or base plate of a test stand.
Spécification
Dimensions Large
Hauteur (métrique) 32 mm
Largeur (métrique) 25 mm