missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-exo,3-exo-dimethanol, 97%

Product Code. 15440218
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440218 25 g 25g
15420218 1 g 1g
15430218 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440218 Supplier Thermo Scientific Alfa Aesar Supplier No. H53422.14
 Restricted Item
Expédition estimée: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 699-95-6 | C9H14O2 | 154.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-95-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00799568
InChI Key IGHHPVIMEQGKNE-ROVKLQMOSA-N
Synonym bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol
PubChem CID 73995587
IUPAC Name [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO

Specifications

Density 1.027
Boiling Point 97°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.523
Quantity 25 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 5-Norbornene-2-exo,3-exo-dimethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.