Learn More
Mecmesin™ Pince-étau 500 N
Code produit 11937989
Encompass
Modifier l'affichage
Click to view available options
Dimensions:
Small
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Dimensions Conditionnement
11937989 Small 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11937989 Fournisseur Mecmesin™ Code fournisseur 432385
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
Ideal for tensile testing below 500 N.
  • Ideal for tensile testing below 500 N.
  • A spring-loaded lever ensures the serrated jaws are easy to open for fast securing and release of samples. As load is applied, the self-tightening action of the wedge jaws ensures the sample is held rigidly without slippage.
  • Fitted with a 10-32 UNF threaded hole and grub-screw for fixing to the base plate of a test stand. When fixing to a loadcell/gauge these wedge grips are often be used with a chain-link assembly to provide a flexible connection.
Spécification
Dimensions Small
Hauteur (métrique) 20 mm
Largeur (métrique) 16 mm
Plage <500 N