Learn More
5Alpha-Cholestan-3Beta-ol-d7, TRC
Code produit 30362667
Change view
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30362667 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30362667 Supplier Toronto Research Chemicals Supplier No. TRCC4317121MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau 5alpha-Cholestan-3beta-ol-d7
CAS 84061-69-8
Formule moléculaire C27H41D7O
Quantité 1 mg
Synonyme (3β,5α)-Cholestan-25,26,26,26,27,27,27-d7-3-ol, (3β,5α)-Cholestan-3-ol-d7, 3β-Hydroxy-5α-cholestane-d7, 3β-Hydroxycholestane-d7, 5α(H)-Cholestan-3β-ol-d7, 5α-Cholestanol-d7, 5α-Dihydrocholesterol-d7, Cholestan-3β-ol-d7, Cholestanol-d7, Dihydrocholesterin-d7, Dihydrocholesterol-d7, Dihydrolathosterol-d7, Lathostanol-d7, NSC 18188-d7, Zymostanol-d7, β-Cholestanol-d7
Formule InChI InChI=1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19?,20-,21-,22-,23+,24-,25-,26-,27+/m0/s1
SMILES O[C@H]1CC[C@@]2(C)[C@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])C(CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])C1
Nom IUPAC (3S,5S,8R,9S,10S,13R,14S,17R)-10,13-dimetil-17-(6-metilheptan-2-il)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahidro-1H-ciclopenta[a]fenantren-3-ol
Poids moléculaire (g/mol) 395.71
Poids de la formule 395.4145