Learn More
STEMCELL Technologies ACCUTASE™
Code produit 15321693
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 ml
500 ml
Conditionnement:
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15321693 100 ml -
17189911 500 ml 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15321693 Fournisseur STEMCELL Technologies Code fournisseur 07920
Cell detachment solution
  • A cell detachment solution of proteolytic and collagenolytic enzymes
  • Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware
  • ACCUTASE™ does not contain mammalian or bacterial-derived products
  • Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic
Spécification
Type de cellule Neural Cells, PSC-Derived, Pluripotent Stem Cells
Quantité 100 ml