Learn More
Acébutolol-d5, TRC
Code produit 30324815
Change view
Click to view available options
Quantité:
10 mg
Conditionnement:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30324815 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324815 Supplier Toronto Research Chemicals Supplier No. TRCA12380210MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau Acebutolol-d5
CAS 1189500-68-2
Notes de qualité de la pureté HPLC
Formule moléculaire C18H23D5N2O4
Quantité 10 mg
Formule InChI InChI=1S/C17H26N2O4/c1-4-6-17(22)19-13-7-8-16(15(9-13)12(3)20)23-11-14(21)10-18-5-2/h7-9,14,18,21H,4-6,10-11H2,1-3H3,(H,19,22)/i10D2,11D2,14D
SMILES [2H]C([2H])(NC(C)C)C([2H])(O)C([2H])([2H])Oc1ccc(NC(=O)CCC)cc1C(=O)C
Nom IUPAC N-[3-acétyl-4-[1,1,2,3,3-pentadeutérin-3-(éthylamino)-2-hydroxypropoxy]phényl]butanamide
Poids moléculaire (g/mol) 341.46
Pourcentage de pureté >95