Learn More
Advanced Instruments Osmolality Linearity Set
For monitoring performance of any osmometer in the range 0-2000 mOsm/kg
Marque:  Advanced Instruments 3LA028
Encompass_Preferred
Code produit. 15168606
  • 199.65€ / 10 pièces
Description
  • Each level of standard supplied in the set should be run and documented to verify the reportable range of patient results
  • Run the linearity set until accurate results obtained or recalibration needed
  • For in vitro diagnostic use with all brands of osmometers