Learn More
Alisol F, MedChemExpress
Code produit 16449138
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16449138 10 mg
16401548 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16449138 Fournisseur Medchem Express Code fournisseur HYN0854
MedChemExpress Alisol F is a triterpene isolated from Alisma orientalis, has immunosuppressive and anti-virus functions. Alisol F exhibits inhibitory activity in vitro on hepatitis B virus (HBV) surface antigen (HBsAg) secretion of the HepG2.2.15 cell line with an IC50 of 0.6 μM.
Alisol F is a triterpene isolated from Alisma orientalis, has immunosuppressive and anti-virus functions. Alisol F exhibits inhibitory activity in vitro on hepatitis B virus (HBV) surface antigen (HBsAg) secretion of the HepG2.2.15 cell line with an IC50 of 0.6 μM.
Spécification
Nom chimique ou matériau Alisol F
Forme physique Solid
CAS 155521-45-2
Couleur Blanc
Quantité 10 mg
Notes de qualité de la pureté Research
Formule moléculaire C30H48O5
À utiliser avec (application) Neuroscience-Neuromodulation
Informations sur la solubilité DMSO : 100 mg/mL (204.62 mM; Need ultrasonic)
SMILES C[C@]1(C(C[C@@H]2O)=C([C@@H]3C)[C@](O[C@@]([C@@H](O)C(C)(O)C)([H])C3)([H])C1)[C@]([C@]2([H])[C@]4(CCC5=O)C)(CC[C@@]4([H])C5(C)C)C
Poids moléculaire (g/mol) 488.7
Poids de la formule 488.7
Pourcentage de pureté 96.2%
Qualité Research