Learn More
Eurofins DiscoverX ALK1 Protein, Active
Description
ALK1 is a type I cell-surface receptor for the TGF-beta superfamily of ligands. It shares with other type I receptors a high degree of similarity in serine-threonine kinase subdomains, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. The encoded protein, sometimes termed ALK1, shares similar domain structures with other closely related ALK or activin receptor-like kinase proteins that form a subfamily of receptor serine/threonine kinases. Mutations in this gene are associated with hemorrhagic telangiectasia type 2, also known as Rendu-Osler-Weber syndrome 2.
GenBank L17075.1
Spécification
Spécification
| Formule | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Poids moléculaire | 45 kDa |
| Quantité | 10 μg |
| Espèces | Human |
| Recombinant | Recombinant |
| Marqueur de protéine | N-terminal 6His |
| Système d’expression | Recombinant enzyme expressed in Sf21 insect cells |
| Forme protéique | Truncated |
| Pureté ou qualité | ≥70% |
| Protéine | ALK1, active |