Learn More
alpha,alpha-Dimethylphenethylformamide, TRC
Code produit 30134631
Change view
Click to view available options
Quantité:
1 g
10 g
Conditionnement:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30134631 10 g 10g
17901045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30134631 Fournisseur Toronto Research Chemicals Code fournisseur TRCD47665010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 52117-13-2
Formule moléculaire C11 H15 N O
Poids moléculaire (g/mol) 177.24
Synonyme Formamide, N-(1,1-dimethyl-2-phenylethyl)-, Formamide, N-(α,α-dimethylphenethyl)- (7CI), N-(1,1-Dimethyl-2-phenylethyl)formamide, 2-Methyl-3-phenyl-2-N-formylaminopropane, N-(α,α-Dimethylphenethyl)formamide, α,α-Dimethylphenethylformamide, N-(2-Methyl-1-phenylpropan-2-yl)formamide
Nom IUPAC N-(2-methyl-1-phenylpropan-2-yl)formamide
SMILES CC(C)(Cc1ccccc1)NC=O
Spécification
Nom chimique ou matériau alpha,alpha-Dimethylphenethylformamide
Notes de qualité de la pureté HPLC
Quantité 10 g
Formule InChI InChI=1S/C11H15NO/c1-11(2,12-9-13)8-10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3,(H,12,13)
Poids de la formule 177.1154
Pourcentage de pureté >95%