Learn More
Bochem™ Cuve à pansements en aluminium
Product Code. 10192691
Change view
Click to view available options
Diamètre (métrique):
80 mm
Unit Size:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diamètre (métrique) Unit Size
10192691 80 mm 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192691 Supplier Bochem™ Supplier No. 8761
Expédition estimée: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Matériau Aluminium
Forme Rond
Type Cuve à pansements
Hauteur (métrique) 20 mm
Diamètre (métrique) 80 mm