Learn More
Tocris Bioscience™ AM 0902
GREENER_CHOICE
Code produit 18759568
Change view
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18759568 10 mg 10mg
18709827 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18759568 Fournisseur Tocris Bioscience™ Code fournisseur 5914/10
Potent and selective TRPA1 antagonist
AM 0902 is a potent and selective TRPA1 antagonist (IC50 = 0.02 μM for human TRPA1). Exhibits no siginificant activity against human TRPV1 or TRPV4, and rat TRPV1, TRPV3 or TRPM8. Exhibits efficacy in a rat pain model. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one
CAS 1883711-97-4
Plage de pourcentage du dosage >98%
Quantité 10 mg
Formule moléculaire C17H15ClN6O2
Poids de la formule Observed MW: 370.79