Learn More
AMG 837 calcium hydrate, MedChemExpress
Code produit 16488567
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16488567 5 mg
16485938 10 mg
16482087 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16488567 Fournisseur Medchem Express Code fournisseur HY13967B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AMG 837 calcium hydrate is a potent, orally bioavailable and partial agonist of GPR40/FFA1. AMG 837 calcium hydrate inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate could enhance insulin secretion and lower glucose levels in rodents.
AMG 837 calcium hydrate is a potent, orally bioavailable and partial agonist of GPR40/FFA1. AMG 837 calcium hydrate inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate could enhance insulin secretion and lower glucose levels in rodents.
Spécification
Nom chimique ou matériau AMG 837 calcium hydrate
CAS 1259389-38-2
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C26H21F3O3.1/2Ca.H2O
Quantité 5 mg
Informations sur la solubilité DMSO : ≥ 42 mg/mL (92.22 mM)
SMILES [O-]C(C[C@H](C#CC)C1=CC=C(C=C1)OCC2=CC=CC(C3=CC=C(C(F)(F)F)C=C3)=C2)=O.[0.5Ca2+].O
Poids moléculaire (g/mol) 455.45
Poids de la formule 455.45
Pourcentage de pureté 97.23%
Qualité Research
Forme physique Solid
À utiliser avec (application) Metabolism-sugar/lipid metabolism