missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ANKK1, GST Tag Recombinant Protein

Code produit 17148745
Change view
Click to view available options
Quantité:
1 mg
10 μg
100 μg
Conditionnement:
100µg
10µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17148745 100 μg 100µg
15990293 10 μg 10µg
17188785 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17148745 Fournisseur Invitrogen™ Code fournisseur A32888

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ANKK1 recombinant human protein (full length) was expressed in insect cells. The recombinant full length ANKK1 can be used in a number of different applications, but has been specifically validated for use with our LanthaScreen™ Kinase binding assay technology to identify novel kinase inhibitors. ANKK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for ANKK1 is NP_848605.1.

ANKK1 serine/threonine kinase is a member of the RIPK family and is closely linked to the DRD2 gene. ANKK1 is closely linked to DRD2 gene on chr 11, and a well studied restriction fragment length polymorphism designated TaqIA, was originally associated with the DRD2 gene, however, later was determined to be located in exon 8 of ANKK1 gene, where it causes a nonconservative amino acid substitution. It is not clear if ANKK1 plays any role in neuropsychiatric disorders previously associated with Taq1A RFLP.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion Q8NFD2
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris HCl with 0.25mM DTT, 0.1mM PMSF, 10mM glutathione, 25% glycerol, 150mM NaCl, 0.1mM EDTA and no preservative; pH 7.5
Identification génétique (Entrez) 255239
Poids moléculaire 110.9 kDa
Nom Human ANKK1, GST Tag
Plage de pH 7.5
Méthode de purification Purified
Quantité 100 μg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Alias de gène 9930020N01Rik; ANKK1; ankyrin repeat and kinase domain containing 1; ankyrin repeat and protein kinase domain-containing protein 1; PKK2; Protein kinase PKK2; sgK288; Sugen kinase 288; X-kinase
Nom usuel ANKK1
Symbole de gène(s) ANKK1
Famille des kinases RIPKs
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine GST-tag
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Conditions d’expédition Dry Ice
Système d’expression Insect cells
Forme protéique Full Length, Recombinant, Active
Forme Liquid
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.