Learn More
R&D Systems™ APE1 Redox Inhibitor, E3330
Product Code. 18696518 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18696518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18696518 Supplier R&D Systems™ Supplier No. NBP14958110mg
  • ONLINE EXCLUSIVE PRICE
    436.73€
    369.60€ / 10mg
    Économisez 67.13€
    15% de réduction
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Product Line Novus
Quantity 10 mg
Type Inhibitor