Learn More
Molecular Devices Aquamax™ 2000 Plate Washer Bundle
Code produit 31935852 Tous les produits Molecular Devices Produits
Modifier l'affichage
Click to view available options
Type de plaques:
96 and 384-well Plates
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de plaques Conditionnement
31935852 96 and 384-well Plates 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 31935852 Fournisseur Molecular Devices Code fournisseur AQUAMAX 2K BUNDLE
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
Règlementaire : Cet article est soumis à éco-contribution.
Automate microplate washing workflows with a complete solution. Bundle supports configurable wash protocols and reliable performance for microplate‐based assays.
  • Provides a complete solution for automated microplate washing workflows
  • Supports multiple wash protocols using configurable fluid inlet systems
  • Enables precise dispense and aspiration control for consistent assay results
  • Supports processing of microplates without cell disruption
  • Supports multiple washing modes including sequential, continuous, and bottom washing
  • Compatible with interchangeable wash heads for different microplate formats
  • Provides flexibility to support current and higher-density microplate applications
  • Designed for use in microplate-based assay workflows across life science applications
TRUSTED_SUSTAINABILITY
Spécification
Flacons Wash Bottle (2 L), Waste Bottle (2 L)
Volume distribué 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
Type de plaques 96 and 384-well Plates
Volume résiduel ≤2 μL/well
Durée de trempage 1 to 600 sec.
Type Plate Washer
Cycles de lavage 1 to 250
Volume de lavage 25 to 3000 μL/well