Learn More
Astemizole, MedChemExpress
Code produit 16441457
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16441457 50 mg
16425248 10 mg
16407967 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 16441457 Fournisseur Medchem Express Code fournisseur HY12532
MedChemExpress Astemizole (R 43512), a second-generation antihistamine drug to diminish allergic symptoms with a long duration of action, is a histamine H1-receptor antagonist, with an IC50 of 4 nM. Astemizole also shows potent hERG K+ channel blocking activity with an IC50 of 0.9 nM. Astemizole has antipruritic effects.
Astemizole (R 43512), a second-generation antihistamine drug to diminish allergic symptoms with a long duration of action, is a histamine H1-receptor antagonist, with an IC50 of 4 nM. Astemizole also shows potent hERG K+ channel blocking activity with an IC50 of 0.9 nM. Astemizole has antipruritic effects.
Spécification
Nom chimique ou matériau Astemizole
CAS 68844-77-9
Couleur Off-White
Notes de qualité de la pureté Research
Formule moléculaire C28H31FN4O
Quantité 50 mg
Synonyme R 43512
Informations sur la solubilité DMSO : 125 mg/mL (272.59 mM; Need ultrasonic)
SMILES COC1=CC=C(CCN2CCC(NC3=NC4=CC=CC=C4N3CC5=CC=C(F)C=C5)CC2)C=C1
Poids moléculaire (g/mol) 458.57
Poids de la formule 458.57
Pourcentage de pureté 98.0%
Qualité Research
Forme physique Solid
À utiliser avec (application) COVID-19-immunoregulation
Afficher plus Afficher moins
Research purposes only
Nom du produit
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.