Learn More
Specac™ Atlas™ Evacuable Pellet Die 13mm
Code produit 12910880
Encompass_Preferred
Modifier l'affichage
Click to view available options
Diamètre (métrique):
13 mm
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Diamètre (métrique) Conditionnement
12910880 13 mm 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12910880 Fournisseur Specac™ Code fournisseur GS03000
Expédition estimée: 11-01-2027
13mm Evacuable Pellet Die used for compacting powdered samples into discs or briquettes with use of a pressing system. Well suited for KBr pellet production for FTIR analysis and XRF pellets. This product comprises of a base, body, plunger, pair of pressing surfaces, extractor ring + O-Ring Kit.
Spécification
Diamètre (métrique) 13 mm