Learn More
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat
Code produit 16462859
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16462859 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16462859 Fournisseur MedChemExpress Code fournisseur HYP1236
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat is a major circulating form of ANP in rats, potently inhibits Angiotensin II (Ang II)-stimulated endothelin-1 secretion in a concentration-dependent manner.
Spécification
Pureté 97.72%
CAS 88898-17-3
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 3062.41 g/mol
Type de produit Peptides
Quantité 1 mg
Conditions d’expédition Room temperature
Synonyme Atrial natriuretic factor (1-28) (rat)