Learn More
BAI1, MedChemExpress
Code produit 16454866
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16454866 100 mg
16453818 10 mg
16436547 5 mg
16430197 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16454866 Fournisseur Medchem Express Code fournisseur HY103269
MedChemExpress BAI1 is a selective and allosteric inhibitor of BAX, an apoptosis regulator. BAI1 directly binds to BAX and allosterically inhibits BAX activation. BAI1 has the potential for the research of diseases mediated by BAX-dependent cell death.
BAI1 is a selective and allosteric inhibitor of BAX, an apoptosis regulator. BAI1 directly binds to BAX and allosterically inhibits BAX activation. BAI1 has the potential for the research of diseases mediated by BAX-dependent cell death.
Spécification
Nom chimique ou matériau BAI1
CAS 335165-68-9
Couleur Off-White
Notes de qualité de la pureté Research
Formule moléculaire C19H21Br2N3O
Quantité 100 mg
Informations sur la solubilité DMSO : 18.67 mg/mL (39.96 mM; Need ultrasonic)
SMILES OC(CN1CCNCC1)CN2C3=C(C4=C2C=CC(Br)=C4)C=C(Br)C=C3
Poids moléculaire (g/mol) 467.2
Poids de la formule 467.2
Pourcentage de pureté 99.73%
Qualité Research
Forme physique Powder
À utiliser avec (application) Cancer-programmed cell death