Learn More
MedChemExpress Balapiravir
Code produit 16433978
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16433978 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16433978 Fournisseur MedChemExpress Code fournisseur HY10443
MedChemExpress Balapiravir (Ro 4588161; R1626) is an orally active prodrug of a nucleoside analogue inhibitor of the RNA-dependent RNA polymerase (RdRp) of HCV (R1479; 4'-Azidocytidine). Balapiravir has anti-HCV activity.
Spécification
Poids moléculaire 494.5 g/mol
Type de produit Reference compound
Contenu et stockage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Conditions d’expédition Room temperature
Pureté 96.97%
Quantité 10 mg
Informations sur la solubilité DMSO : ≥ 100 mg/mL (202.22 mM)
À utiliser avec (application) COVID-19-anti-virus
CAS 690270-29-2
Synonyme Ro 4588161∣ R1626