Learn More
MedChemExpress Balapiravir
Code produit 16466687
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16466687 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16466687 Fournisseur MedChemExpress Code fournisseur HY10443
MedChemExpress Balapiravir (Ro 4588161; R1626) is an orally active prodrug of a nucleoside analogue inhibitor of the RNA-dependent RNA polymerase (RdRp) of HCV (R1479; 4'-Azidocytidine). Balapiravir has anti-HCV activity.
Spécification
Poids moléculaire 494.5 g/mol
Type de produit Reference compound
Contenu et stockage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Conditions d’expédition Room temperature
Pureté 96.97%
Quantité 5 mg
Informations sur la solubilité DMSO : ≥ 100 mg/mL (202.22 mM)
À utiliser avec (application) COVID-19-anti-virus
CAS 690270-29-2
Synonyme Ro 4588161∣ R1626