Learn More
BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Code produit 12927746
Modifier l'affichage
Click to view available options
Calibre d’aiguille:
23 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Calibre d’aiguille
12927746 23 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12927746 Fournisseur BD Code fournisseur 367324
Expédition estimée: 18-01-2027
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
  • Ease of use reduces training requirements and improves activation compliance
  • In-vein activation reduces the risk exposure to a contaminated needle
  • Easy activation without additional patient discomfort
  • Ideal for use in high-risk environments No luer adapter
TRUSTED_SUSTAINABILITY
Spécification
Type de produit Push Button Blood Collection Set
Calibre d’aiguille 23 g
Longueur (impériale) de l’aiguille 3/4 in.
Longueur (impériale) du tube 12 in