Learn More
BET-BAY 002, MedChemExpress
Product Code. 16481377
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
16481377 50 mg
16437877 5 mg
16405866 100 mg
16405158 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481377 Supplier Medchem Express Supplier No. HY12421
 Article restreint
Expédition estimée: 10-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress BET-BAY 002 is a potent BET inhibitor; shows efficacy in a multiple myeloma model.
BET-BAY 002 is a potent BET inhibitor
Specifications
Nom chimique ou matériau BET-BAY 002
Forme physique Solid
CAS 1588521-78-1
Couleur Blanc
Quantité 50 mg
Notes de qualité de la pureté Research
Formule moléculaire C22H18ClN5O
À utiliser avec (application) Cancer-programmed cell death
Informations sur la solubilité DMSO : ≥ 50 mg/mL (123.81 mM)
SMILES CC1=NN=C(C[C@H]2C3=NN=C(C)N3C4=CC=CC=C4C(C5=CC=C(Cl)C=C5)=C2)O1
Poids moléculaire (g/mol) 403.86
Poids de la formule 403.86
Pourcentage de pureté 99.52%
Qualité Research