Learn More
BETd-246, MedChemExpress
Code produit 16454858
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16454858 10 mg
16447607 5 mg
16433266 25 mg
16421117 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16454858 Fournisseur Medchem Express Code fournisseur HY115568
MedChemExpress BETd-246 is a second-generation and PROTAC-based BET bromodomain (BRD) inhibitor connected by ligands for Cereblon and BET, exhibiting superior selectivity, potency and antitumor activity.
BETd-246 is a second-generation and PROTAC-based BET bromodomain (BRD) inhibitor connected by ligands for Cereblon and BET, exhibiting superior selectivity, potency and antitumor activity.
Spécification
Nom chimique ou matériau BETd-246
CAS 2140289-17-2
Couleur Jaune
Notes de qualité de la pureté Research
Formule moléculaire C48H55N11O10
Quantité 10 mg
Informations sur la solubilité DMSO : 200 mg/mL (211.41 mM; Need ultrasonic)
SMILES O=C(C1=NC(NC2=CC(C3CC3)=NN2CC)=C4C(NC5=C4C=C(OC)C(C6=C(C)ON=C6C)=C5)=N1)NCCCOCCOCCOCCCNC7=CC=CC(C(N8C(CC9)C(NC9=O)=O)=O)=C7C8=O
Poids moléculaire (g/mol) 946.02
Poids de la formule 946.02
Pourcentage de pureté 98.04%
Qualité Research
Forme physique Solid
À utiliser avec (application) Neuroscience-Neuromodulation