Learn More
Tocris Bioscience™ BIMU 8
GREENER_CHOICE
Code produit 18717246
Change view
Click to view available options
Quantité:
5 mg
25 mg
Conditionnement:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18717246 25 mg 25mg
18759206 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18717246 Fournisseur Tocris Bioscience™ Code fournisseur 4374/25
Potent 5-HT4 agonist
BIMU 8 is a potent 5-HT4 receptor full agonist (KD = 77 nM and EC50 = 18 nM for wild type 5-HT4 receptors). Binds to receptors in a mechanism different to that of the endogenous ligand 5-HT.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 2,3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride
CAS 134296-40-5
Plage de pourcentage du dosage >98%
Quantité 25 mg
Formule moléculaire C19H26N4O2.HCl
Poids de la formule Observed MW: 378.9