Learn More
Thermo Scientific™ BioProcess Container (BPC), 50 L, for HyPerforma™ Single-Use Mixer DS 300, Bottom drain, irradiated
Code produit 30040288 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Quantité:
Unité
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30040288 Unité 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30040288 Fournisseur Thermo Scientific™ Code fournisseur SH30946.01
 Article restreint
Expédition estimée: 02-03-2027
The 50 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.
The 50 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.
Key features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
  • All standard BPCs and liners are designed to fit 'out-of-the-box'
  • Available in volume options of 50, 100, and 200 L
  • Applicable to open- and closed-top mixing applications
  • Powder-liquid and liquid-liquid mixing
  • World class CX5-14 and CX3-9 film
  • Adaptable to multiple upstream process applications in media and buffer prep
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma SIngle-Use Mixer DS 300 drums
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single use
  • High-throughput mixing and storage
  • Additional configurations are available to suit your application requirements.
Accessory
  • Reusable Bearing Hub (Cat No. SV5017777)
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 50 L
Conditionnement externe Les boîtes sont placées dans des cartons solides avec identification du produit et du lot
Conditionnement interne protecteur Conditionnés et scellés en deux couches externes indépendantes dans la zone de Classe 10 000 / ISO Classe 7
Quantité Unité
Gamme de produits HyPerforma