Learn More
STEMCELL Technologies Blunt-End Needles, 16 Gauge
Code produit 16625655
Encompass_Preferred
Modifier l'affichage
Click to view available options
Calibre d’aiguille:
16 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Calibre d’aiguille
16625655 16 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16625655 Fournisseur STEMCELL Technologies Code fournisseur 28110
 Article restreint
Expédition estimée: 22-12-2026
Sterile, 16 gauge, metal luer lock hub, 1.5 in. long
  • Sterile 16 gauge needles, 1.5 inches long with a metal luer lock hub are ideal for safe and accurate handling of methylcellulose-based medium (e.g. MethoCult™)
  • The blunt end reduces the risk of needle-stick injuries
  • Product is manufactured from non-pyrogenic materials
Spécification
Calibre d’aiguille 16 g
À utiliser avec (application) Colony Assay
Quantité 100/Pk.