Learn More
R&D Systems™ BromoCatch™ Expression Plasmid (N-term, pTK, Puro)
Code produit 31900382
Modifier l'affichage
Click to view available options
Quantité:
50 μg
Conditionnement:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
31900382 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 31900382 Fournisseur R&D Systems™ Code fournisseur RDELBC01050
 Article restreint
Expédition estimée: 02-11-2026
This plasmid enables expression of proteins fused to the BromoCatch™ tag, a covalent self-labeling tag derived from a BET bromodomain.
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits R&D Systems
Quantité 50 μg