Learn More
Unity Lab Services™ Calibration Certificate Shore A Durometer
Description
- Verification according to standard: NF T 51-123
- Control of indentors on the profile projector
- Check springs with a balance
Specifications
Specifications
| For Use With (Equipment) | Shore A Durometers |
| Duration | 1 Day |
| Product Type | Calibration Certificate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?