Learn More
DiscoverX cAMP Hunter™ CHO-K1 LHCGR Bioassay Kit
Description
The cAMP Hunter CHO-K1 LHCGR Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay asseses ligand (e.g. Peptide YY, Human) based on activation of LHCGR GPCR activity via detection of cAMP. The included cell line overexpresses ProLink-tagged LHCGR and EA-tagged . Activation of LHCGR stimulates the recruitment of and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Spécification
Spécification
| Entrée du dosage | Intracellular cAMP level |
| À utiliser avec (application) | Assay-Ready Bioassay Kits |
| Quantité | 1 000 ddp (10 x 96 puits) |
| Lignée cellulaire | CHO-K1 |
| Cible | LHCGR |