Learn More
Thermo Scientific™ Capit-All Flex Cap Driver Cassettes
Code produit 31872965 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Tubes de stockage compatibles:
Azenta 96 Format External Thread
Azenta 96 Format Internal Thread
Matrix and Nunc tubes in 96 or 48 formats
Conditionnement:
1 pièce
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Tubes de stockage compatibles Conditionnement
31872965 Azenta 96 Format External Thread 1 pièce
31872964 Azenta 96 Format Internal Thread 1 pièce
30274494 Matrix and Nunc tubes in 96 or 48 formats 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 31872965 Fournisseur Thermo Scientific™ Code fournisseur 4130FDXE
 Article restreint
Expédition estimée: 29-12-2026
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
  • Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
  • Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
  • Replace after 10,000 cycles
TRUSTED_SUSTAINABILITY
Spécification
Tubes de stockage compatibles Azenta 96 Format External Thread
Format de débouchage 96-Channel
Type Decapping Head