Learn More
MedChemExpress CEP dipeptide 1
Code produit 16467196
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16467196 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16467196 Fournisseur MedChemExpress Code fournisseur HY16959
MedChemExpress CEP dipeptide 1 is a CEP dipeptide with potent angiogenic activity; mediators of age-related macular degeneration (AMD).
Spécification
Pureté 98.0%
CAS 816432-15-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Poids moléculaire 381.42 g/mol
Type de produit Peptides
Quantité 100 mg
Conditions d’expédition Room temperature
Solubilité H2O : 10 mg/mL (26.22 mM∣Need ultrasonic)