Learn More
MedChemExpress CEP dipeptide 1
Code produit 16479747
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16479747 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16479747 Fournisseur MedChemExpress Code fournisseur HY16959
MedChemExpress CEP dipeptide 1 is a CEP dipeptide with potent angiogenic activity; mediators of age-related macular degeneration (AMD).
Spécification
Pureté 98.0%
CAS 816432-15-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Poids moléculaire 381.42 g/mol
Type de produit Peptides
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité H2O : 10 mg/mL (26.22 mM∣Need ultrasonic)