missing translation for 'onlineSavingsMsg'
Learn More

Chloroform Stabilised With Ethanol, Analytical Grade, Penta Chemicals

Code produit 30020508
Modifier l'affichage
Click to view available options
Quantité:
1 L
2.5 L
Conditionnement:
1L
2.50L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30020508 1 L 1L
30020516 2.5 L 2.50L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire: Ce produit contient une substance répertoriée dans l'annexe XVII du règle ment REACH (CE) N° 1907/2006, tout client doit remplir une déclaration avant la livraison.
Code produit 30020508 Fournisseur PENTA CHEMICALS Code fournisseur 1713011000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire: Ce produit contient une substance répertoriée dans l'annexe XVII du règle ment REACH (CE) N° 1907/2006, tout client doit remplir une déclaration avant la livraison.

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Identifiants chimiques

CAS 67-66-3
Poids moléculaire (g/mol) 119.38

Spécification

Nom chimique ou matériau Chloroform
Quantité 1 L
Conditionnement Flacon en verre
Pourcentage de pureté ≥99.8%
Qualité Analytique
compliance-icons compliance-icons
Identifiant produit
  • Chloroform stabilised with ethanol
Mention d'avertissement
  • Dangers
Catégorie de danger
  • Toxicité aiguë Catégorie 3
  • Toxicité aiguë Catégorie 4
  • Cancérogénicité Catégorie 2
  • Lésion oculaire grave/irritation oculaire Catégorie 2
  • Toxicité pour la reproduction Catégorie 2
  • Corrosion cutanée/irritation cutanée Catégorie 2
  • Toxicité spécifique pour certains organes cibles à la suite d'une exposition répétée Catégorie 1
  • Toxicité spécifique pour certains organes cibles Catégorie 3
Mention de danger
  • H302-Nocif en cas d'ingestion.
  • H315-Provoque une irritation cutanée.
  • H319-Provoque une sévère irritation des yeux.
  • H331-Toxique par inhalation.
  • H336-Peut provoquer somnolence ou vertiges.
  • H351-Susceptible de provoquer le cancer.
  • H361d-Susceptible de nuire au foetus.
  • H372-Risque avéré d'effets graves pour les organes.
Conseils de prudence
  • P261-Éviter de respirer les poussières/fumées/gaz/brouillards/vapeurs/ aérosols.
  • P280-Porter des gants de protection/des vêtements de protection/un équipement de protection des yeux/ du visage.
  • P305+P351+P338-EN CAS DE CONTACT AVEC LES YEUX: rincer avec précaution à l'eau pendant plusieurs minutes. Enlever les lentilles de contact si la victime en porte et si elles peuvent être facilement enlevées. Continuer à rincer.
  • P311-Appeler un CENTRE ANTIPOISON/un médecin/ .
Informations supplémentaires
  • MIXTURE LIST-Contain: Chloroform, Ethanol
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.