Learn More
Tocris Bioscience™ Ciclopirox
GREENER_CHOICE
Code produit 18773198
Change view
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18773198 50 mg 50mg
18771108 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18773198 Fournisseur Tocris Bioscience™ Code fournisseur 6384/50
Pan-histone demethylase inhibitor
Ciclopirox is a pan-histone demethylase inhibitor. Exhibits no significant effect on histone deacetylases. Inhibits MYC-signaling and proliferation of neuroblastoma cells. Promotes Cdc25A-degradation in cancer cells. Antifungal.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 6-Cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone
CAS 29342-05-0
Plage de pourcentage du dosage >99%
Quantité 50 mg
Formule moléculaire C12H17NO2
Poids de la formule Observed MW: 207.27