Learn More
Ciclopirox, MedChemExpress
Code produit 16448316
Encompass_Preferred
Change view
Click to view available options
Quantité:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16448316 100 mg
16434367 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16448316 Fournisseur Medchem Express Code fournisseur HYB0450
MedChemExpress Ciclopirox (HOE296b) is a synthetic antifungal agent that can be used for superficial mycoses reseaech. Ciclopirox olamine has a very broad spectrum of activity and inhibits dermatophytes, yeasts, molds, and many Gram-positive and Gram-negative species pathogenic.
Ciclopirox (HOE296b) is a synthetic antifungal agent that can be used for superficial mycoses reseaech. Ciclopirox olamine has a very broad spectrum of activity and inhibits dermatophytes, yeasts, molds, and many Gram-positive and Gram-negative species pathogenic.
Spécification
Nom chimique ou matériau Ciclopirox
Forme physique Solid
CAS 29342-05-0
Couleur Off-White
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C12H17NO2
À utiliser avec (application) COVID-19-immunoregulation
Synonyme HOE296b
Informations sur la solubilité DMSO : 100 mg/mL (482.46 mM; Need ultrasonic)
SMILES CC(C=C(C1CCCCC1)N2O)=CC2=O
Poids moléculaire (g/mol) 207.27
Poids de la formule 207.27
Pourcentage de pureté 99.75%
Qualité Research