Learn More
Ciprofibrate, MedChemExpress
Code produit 16498406
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16498406 100 mg
16432196 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16498406 Fournisseur Medchem Express Code fournisseur HYB0664
MedChemExpress Ciprofibrate (Win35833) is a potent peroxisome proliferator and increases the phosphorylation level of the PPARalpha. Ciprofibrate acts as an orally active hypolipidaemic agent and can be used for the research of primary hyperlipidaemias.
Ciprofibrate (Win35833) is a potent peroxisome proliferator and increases the phosphorylation level of the PPARalpha. Ciprofibrate acts as an orally active hypolipidaemic agent and can be used for the research of primary hyperlipidaemias.
Spécification
Nom chimique ou matériau Ciprofibrate
CAS 52214-84-3
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C13H14Cl2O3
Quantité 100 mg
Synonyme Win35833
Informations sur la solubilité DMSO : ≥ 100 mg/mL (345.84 mM)
SMILES CC(C)(OC1=CC=C(C2C(Cl)(Cl)C2)C=C1)C(O)=O
Poids moléculaire (g/mol) 289.15
Poids de la formule 289.15
Pourcentage de pureté 99.79%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death