Learn More
Cobimetinib racemate, MedChemExpress
Product Code. 16485538
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
16485538 10 mg
16481707 50 mg
16458247 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485538 Supplier Medchem Express Supplier No. HY13078
This item is not returnable. View return policy
MedChemExpress Cobimetinib racemate (GDC-0973 racemate; XL518 racemate) is the racemate of Cobimetinib. Cobimetinib is a potent and selective MEK inhibitor.
Cobimetinib racemate (GDC-0973 racemate
Specifications
Nom chimique ou matériau Cobimetinib racemate
CAS 934662-91-6
Couleur Off-White
Notes de qualité de la pureté Research
Formule moléculaire C21H21F3IN3O2
Quantité 10 mg
Synonyme GDC-0973 racemateXL518 racemate
Informations sur la solubilité DMSO : ≥ 40 mg/mL (75.29 mM)
SMILES O=C(C1=CC=C(F)C(F)=C1NC2=CC=C(I)C=C2F)N3CC(C4NCCCC4)(O)C3
Poids moléculaire (g/mol) 531.31
Poids de la formule 531.31
Pourcentage de pureté 95.12%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease