Learn More
Masterflex Cole-Parmer Acrylic Flowmeter, 50 mm Scale for Air, 0.1-1 LPM
Marque:  Masterflex 32457-42
Cole-Parmer Acrylic Flowmeter, 50 mm Scale for Air, 0.1-1 LPM
Code produit. 15531812
  • 103.00€ / 1 pièces
Il en reste null
Expédition estimée: 22-07-2026