Learn More
Kern Sohn Microscope à lumière transmise OBL
Code produit 17399521
Encompass_Preferred
Modifier l'affichage
Click to view available options
Type de tête:
Binoculaire
Trinoculaire
Conditionnement:
1 pièce
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de tête Conditionnement
17399521 Trinoculaire 1 pièce
17389521 Binoculaire 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Règlementaire : Cet article est soumis à éco-contribution.
Code produit 17399521 Fournisseur Kern Sohn Code fournisseur OBL 137
 Article restreint
Expédition estimée: 24-11-2026
Règlementaire : Cet article est soumis à éco-contribution.
The flexible laboratory assistant with infinity optical system and fixed, pre-centred Koehler illumination.
  • The OBL series stands out through its infinity optical unit and is therefore ideally suited for all demanding transmitted illumination applications. The robust and ergonomic stand base guarantees safe and comfortable working
  • Strong and continuously dimmable 3 W-LED illumination
  • The fixed, pre-centred and focusable 1.25 Abbe condenser with aperture diaphragm and field diaphragm gives you a simplified Koehler illumination, without having to move the centre.
  • The large mechanical stage and its specimen holder holds up to two samples at the same time and is quick and easy to focus using a coaxial coarse and fine focusing knob on both sides.
  • A large selection of eyepieces, objectives and colour filters as well as a darkfield condenser, a simple polarising unit, different phase contrast kits through to HBO and LED fluorescence units are available to you as accessories
  • A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
  • Scope of application: haematology, urology, gynaecology, dermatology, pathology, microbiology and parasitology, immunology, oncology, entomology, Veterinary, water analysis, breweries
  • Applications/Samples: Translucent, thin, low-contrast, challenging samples (e.g. living mammal cells, bacteria, tissue)
Spécification
Type de tête Trinoculaire
Type d’éclairage LED
Pouvoir grossissant 4X, 10X, 40X, 100X
Tension 230 V