Learn More
MedChemExpress Crustacean Cardioactive Peptide (CCAP)
Code produit 16429468
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16429468 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16429468 Fournisseur MedChemExpress Code fournisseur HYP0303
MedChemExpress Crustacean Cardioactive Peptide (CCAP) is a highly conserved, amidated cyclic nonapeptide, first isolated from the pericardial organs of the shore crab Carcinus maenas, where it has a role in regulating heartbeat; Crustacean Cardioactive Peptide (CCAP) also modulates the neuronal activity in other arthropods.
Spécification
Pureté 99.66%
CAS 309247-84-5
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 959.1 g/mol
Type de produit Natural Products
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 50 mg/mL (52.13 mM∣Need ultrasonic) ∣H2O : 0.67 mg/mL (0.70 mM∣Need ultrasonic)