Learn More
R&D Systems™ CryoDefend-Cell Lines (5 x 10 mL)
Code produit 18738857
Modifier l'affichage
Click to view available options
Quantité:
1 kit
Conditionnement:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18738857 1 kit 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18738857 Fournisseur R&D Systems™ Code fournisseur CCM019
 Article restreint
Expédition estimée: 16-12-2026
Includes
5 x 10mL vials CryoDefend-Stem Cells Media
Media for defined, protein-free cryopreservation of cultured cells
Cryopreservation can be used to maintain cells at a low passage number and to reduce the risk of contamination by microbes and other cell types. The viability of the recovered cells depends on cell density, the type of cryoprotectant used, the method of cooling, and the amount of the cryoprotectant that remains in the culture after the cells have been thawed.
Contains a defined serum substitute as well as an optimized concentration of a cryopreservative that increases the recovery and viability of healthy cells compared to conventional freezing media
This product contains
DMSO and is ready for use
Reduces experimental variability by using fully defined media
Suitable for all cell types
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits R&D Systems
Quantité 1 kit
Contenu et stockage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.