Learn More
Gibco™ CTS™ DynaCellect™ Bead Removal Kit
Code produit 18239032 Tous les produits Gibco Produits
Modifier l'affichage
Click to view available options
Quantité:
5 kits
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18239032 5 kits 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18239032 Fournisseur Gibco™ Code fournisseur A52301
Valorisez le développement de vos procédés
Utilisez le code promo 27978
Afficher toutes les promotions
 Article restreint
Expédition estimée: 03-12-2026
The Gibco CTS™ DynaCellect™ Bead Removal Kit is designed for depletion and magnetic bead removal on the Gibco CTS™ DynaCellect™ Magnetic Separation System. Design consists of a bead removal chamber, 4 weldable tubes for input and output bags, filters, a cassette, and tube organizer.
The CTS DynaCellect Bead Removal Kit is a single-use, fit-for-purpose kit designed specifically for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal. DynaCellect can help acheive high cell purity, recovery, and viability for any cells of interest.
Characteristics of the CTS DynaCellect Bead Removal Kit include:
• Weldable tube material: 4 x DEHP-free PVC
• Chamber: serpentine design for continuous flow
This kit and the CTS DynaCellect Cell Isolation Kit enable a range of cell processing applications such as isolation, depletion, and bead removal. They offer flexibility for process development and can be customized to support commercial scale manufacture.
Each kit has input/output sterile weldable tubes for flexible setup configurations. They are compatible with sterile welding and are gamma sterilized.
TRUSTED_SUSTAINABILITY
Spécification
Type de produit Bead Removal Kit
Quantité 5 kits
À des fins de recherche ou de fabrication de produits à base de cellules, de gènes ou de tissus. MISE EN GARDE : Non destiné à être directement administré à des humains ou des animaux.