Learn More
R&DSystems™ Cultrex Reduced Growth Factor Basement Membrane Extract, Type 2, Select
Code produit 18412170
Modifier l'affichage
Click to view available options
Quantité:
1 ml
5 ml
Conditionnement:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18412170 1 ml 1mL
18798826 5 ml 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18412170 Fournisseur R&DSystems™ Code fournisseur 353600102
Please to purchase this item. Need a web account? Register with us today!
For Robust Organoid Cultures - Reduced Growth Factor Basement Membrane Extract
/////Qualified for use in organoid cell culture/////Commonly used robust and established organoid systems/////Reduced growth factor formulation provides a more defined culture system/////Quality controlled for performance consistency
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Quantité 1 ml
Espèces Mouse