Learn More
R&D Systems™ Cultrex Stem Cell Qualified Laminin I
Code produit 18716716
Modifier l'affichage
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18716716 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18716716 Fournisseur R&D Systems™ Code fournisseur 340001003
  • ONLINE EXCLUSIVE PRICE
    292.17€
    260.40€ / 1mg
    Économisez 31.77€
    10% de réduction
 Article restreint
Expédition estimée: 20-11-2026
R&D Systems™ Cultrex Stem Cell Qualified Laminin I provides a functionally defined and effective feeder-free surface for the attachment and maintenance of embryonic stem cells in a pluripotent state, thereby enabling its use for growth promotion or study of stem cell differentiation.
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits R&D Systems
Quantité 1 mg
Contenu et stockage Store at -80°C in a manual defrost freezer